Learn More
TARGETMOL CHEMICALS INC ALECTINIB HYDROCHLORIDE 100MG
SDP
Catalog No. 502579081
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 50 mg 500 mg and bulk. Please contact Fisher for quotes. Alectinib hydrochloride (RO5424802 Hydrochloride) is a selective and orally available inhibitor of ALK( IC50 1.9 nM)purity: 99%
Catalog No. 50-257-9081 Supplier TARGETMOL CHEMICALS INC Supplier No. T8387100MG
Also available in 5 mg 10 mg 25 mg 50 mg 500 mg and bulk. Please contact Fisher for quotes. Alectinib hydrochloride (RO5424802 Hydrochloride) is a selective and orally available inhibitor of ALK( IC50 1.9 nM)purity: 99%