Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Alectinib (Hydrochloride) | 1256589-74-8 | 99.9% | 100 MG
SDP

Catalog No. 5000338593
Encompass_Preferred

Alectinib Hydrochloride is a potent, selective, and orally available ALK inhibitor with an IC50 of 1.9 nM and a Kd value of 2.4 nM (in an ATP-competitive manner). It effectively inhibits ALK F1174L and ALK R1275Q with IC50s of 1 nM and 3.5 nM, respectively, and demonstrates effective central nervous system (CNS) penetration.

  • Potent, selective, and orally available ALK inhibitor
  • Effective central nervous system (CNS) penetration
  • Molecular formula: C30H35ClN4O2
  • Molecular weight: 519.08
  • Appearance: White to off-white solid
  • Recommended storage at 4°C, sealed and away from moisture

Catalog No. 50-003-38593 Supplier Medchemexpress LLC Supplier No. HY13011A100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Alectinib Hydrochloride is a potent, selective, and orally available ALK inhibitor with an IC50 of 1.9 nM and a Kd value of 2.4 nM (in an ATP-competitive manner). It effectively inhibits ALK F1174L and ALK R1275Q with IC50s of 1 nM and 3.5 nM, respectively, and demonstrates effective central nervous system (CNS) penetration.

  • Potent, selective, and orally available ALK inhibitor
  • Effective central nervous system (CNS) penetration
  • Molecular formula: C30H35ClN4O2
  • Molecular weight: 519.08
  • Appearance: White to off-white solid
  • Recommended storage at 4°C, sealed and away from moisture

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.