Learn More
Medchemexpress LLC Alectinib Hydrochloride | 1256589-74-8 | 99.9% | 200 MG
SDP
Catalog No. 5000338586
Encompass_Preferred
Alectinib (CH5424802; RO5424802; RG7853) Hydrochloride is a potent, selective, and orally available ALK inhibitor. It has an IC50 of 1.9 nM and a Kd value of 2.4 nM (in an ATP-competitive manner). It also inhibits ALK F1174L and ALK R1275Q with IC50s of 1 nM and 3.5 nM, respectively. Alectinib demonstrates effective central nervous system (CNS) penetration.
  • Potent and selective ALK inhibitor
  • Orally available
  • Effective central nervous system (CNS) penetration
  • Inhibits ALK F1174L and ALK R1275Q
Catalog No. 50-003-38586 Supplier Medchemexpress LLC Supplier No. HY13011A200MG
Alectinib (CH5424802; RO5424802; RG7853) Hydrochloride is a potent, selective, and orally available ALK inhibitor. It has an IC50 of 1.9 nM and a Kd value of 2.4 nM (in an ATP-competitive manner). It also inhibits ALK F1174L and ALK R1275Q with IC50s of 1 nM and 3.5 nM, respectively. Alectinib demonstrates effective central nervous system (CNS) penetration.
  • Potent and selective ALK inhibitor
  • Orally available
  • Effective central nervous system (CNS) penetration
  • Inhibits ALK F1174L and ALK R1275Q