Learn More
Medchemexpress LLC Alectinib hydrochloride | 1256589-74-8 | 99.9% | 25 MG
SDP
Catalog No. 5000341611
Encompass_Preferred
Alectinib hydrochloride is a potent, selective, and orally available ALK inhibitor. It demonstrates effective central nervous system (CNS) penetration.
  • Potent and selective ALK inhibitor
  • Orally available
  • Inhibits ALK F1174L and ALK R1275Q
  • Effective central nervous system (CNS) penetration
  • Reduces cell activity in a dose-dependent manner
  • Leads to dose-dependent tumor growth inhibition and regression
Catalog No. 50-003-41611 Supplier Medchemexpress LLC Supplier No. 25MGHY13011A
Alectinib hydrochloride is a potent, selective, and orally available ALK inhibitor. It demonstrates effective central nervous system (CNS) penetration.
  • Potent and selective ALK inhibitor
  • Orally available
  • Inhibits ALK F1174L and ALK R1275Q
  • Effective central nervous system (CNS) penetration
  • Reduces cell activity in a dose-dependent manner
  • Leads to dose-dependent tumor growth inhibition and regression