Learn More
Medchemexpress LLC Alectinib hydrochloride | 1256589-74-8 | MFCD27987893 | 99.9% | C30H35ClN4O2 | 10MG
SDP
Catalog No. 5000260165
Encompass_Preferred
Alectinib hydrochloride is the hydrochloride salt of alectinib, a potent and selective anaplastic lymphoma kinase (ALK) inhibitor. It is supplied as a research-grade analytical standard for biochemical and cell-based assays and includes documentation to support analytical verification. Intended for research use only.
  • High purity (99.9%) for analytical applications.
  • Verified identity with CAS 1256589-74-8.
  • Available in multiple small pack sizes for method development.
  • Soluble in common organic solvents for assay preparation.
  • Supplied with COA and datasheet for traceability.
Catalog No. 50-002-60165 Supplier Medchemexpress LLC Supplier No. HY13011A10MG
Alectinib hydrochloride is the hydrochloride salt of alectinib, a potent and selective anaplastic lymphoma kinase (ALK) inhibitor. It is supplied as a research-grade analytical standard for biochemical and cell-based assays and includes documentation to support analytical verification. Intended for research use only.
  • High purity (99.9%) for analytical applications.
  • Verified identity with CAS 1256589-74-8.
  • Available in multiple small pack sizes for method development.
  • Soluble in common organic solvents for assay preparation.
  • Supplied with COA and datasheet for traceability.