Learn More
Enzo Life Sciences Alectinib. hydrochloride (500mg). CAS: 1256589-74-8
SDP
Catalog No. 502013542
Encompass_Preferred
Alectinib hydrochloride is a selective anaplastic lymphoma kinase (ALK) inhibitor leading to inhibition of tumor cell proliferation and induction of cell death.  Purity: ≥99% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: -20°C.
Catalog No. 50-201-3542 Supplier Enzo Life Sciences Supplier No. ENZCHM2130500
Alectinib hydrochloride is a selective anaplastic lymphoma kinase (ALK) inhibitor leading to inhibition of tumor cell proliferation and induction of cell death.  Purity: ≥99% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: -20°C.