Learn More
New England Biolabs, Inc. AleI-v2 – 2500 units
SDP
Catalog No. 501959364
Encompass
AleI-v2 is an engineered and improved version of AleI that recognizes the sequence CACNN^NNGTG.
Catalog No. 50-195-9364 Supplier New England Biolabs, Inc. Supplier No. R0685L
AleI-v2 is an engineered and improved version of AleI that recognizes the sequence CACNN^NNGTG.