Learn More
New England Biolabs, Inc. AleI-v2 – 500 units
SDP
Catalog No. NC1595347
Encompass
AleI-v2 is an engineered and improved version of AleI that recognizes the sequence CACNN^NNGTG.
Catalog No. NC1595347 Supplier New England Biolabs, Inc. Supplier No. R0685S
AleI-v2 is an engineered and improved version of AleI that recognizes the sequence CACNN^NNGTG.