Learn More
Medchemexpress LLC HY-P7003 10ug Medchemexpress, BAFF/TNFSF13B (CHO-expressed), Human CAS: Purity:>98%
SDP
Catalog No. 501970551
Encompass_Preferred
Medchemexpress, HY-11101 100mg Alendronate (sodium hydrate) CAS:121268-17-5 Alendronate (sodium hydrate) is a farnesyl diphosphate synthase inhibitor with IC50 of 460 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0551 Supplier Medchemexpress LLC Supplier No. HYP700310UG
Medchemexpress, HY-11101 100mg Alendronate (sodium hydrate) CAS:121268-17-5 Alendronate (sodium hydrate) is a farnesyl diphosphate synthase inhibitor with IC50 of 460 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.