Learn More
Cayman Chemical Alendronate (sodium hydrate), Purity: ≥95%, CAS: 121268-17-5, 100 mg
SDP
Catalog No. 502955143
Encompass_Preferred
An FPP synthase inhibitor (IC50 = 50 nM for the recombinant human enzyme); inhibits bone resorption in a pit formation assay using isolated rat osteoclasts (IC50 = 2 nM); reduces angiotensin II-induced collagen I synthesis in isolated rat cardiac fibroblasts at 3 and 10 µM; prevents femoral bone mineral density loss in ovariectomized rats at 5 mg/kg
Catalog No. 50-295-5143 Supplier Cayman Chemical Supplier No. 13642100 MG
An FPP synthase inhibitor (IC50 = 50 nM for the recombinant human enzyme); inhibits bone resorption in a pit formation assay using isolated rat osteoclasts (IC50 = 2 nM); reduces angiotensin II-induced collagen I synthesis in isolated rat cardiac fibroblasts at 3 and 10 µM; prevents femoral bone mineral density loss in ovariectomized rats at 5 mg/kg