Learn More
Apexbio Technology LLC Alendronic acid 66376-36-1 200mg
SDP
Catalog No. 5000090379
Encompass_Preferred
Alendronic acid is a small-molecule inhibitor targeting farnesyl diphosphate synthase (FDPS) It is designed to inhibit FDPS activity thereby disrupting protein prenylation pathways required for osteoclast functionality and regulating osteoclast-mediated bone resorption Alendronic acid exerts its biological activity primarily through inhibition of FDPS which suppresses osteoclast activity and associated bone mineral degradation Based on these pharmacological properties alendronic acid holds research potential in the investigation of bone remodeling mechanisms particularly in the contexts of postmenopausal osteoporosis Paget s disease and tumor-related hypercalcemia
Catalog No. 50-000-90379 Supplier Apexbio Technology LLC Supplier No. C6689200
Alendronic acid is a small-molecule inhibitor targeting farnesyl diphosphate synthase (FDPS) It is designed to inhibit FDPS activity thereby disrupting protein prenylation pathways required for osteoclast functionality and regulating osteoclast-mediated bone resorption Alendronic acid exerts its biological activity primarily through inhibition of FDPS which suppresses osteoclast activity and associated bone mineral degradation Based on these pharmacological properties alendronic acid holds research potential in the investigation of bone remodeling mechanisms particularly in the contexts of postmenopausal osteoporosis Paget s disease and tumor-related hypercalcemia