Learn More
Medchemexpress LLC Alendronic acid-d6 | 1035437-39-8 | 98.0% | 255.13 | C4H7D6NO7P2 | 10 MG
SDP
Catalog No. 5000297090
Encompass_Preferred
Alendronic acid-d6 is the deuterium-labeled analogue of alendronic acid supplied as a solid stable-isotope reagent for use as a tracer and internal standard in quantitative analytical methods such as NMR, GC-MS, and LC-MS. Purity 98.0%; molecular weight 255.13; molecular formula C4H7D6NO7P2. Storage: powder at -20°C (up to 3 years) or 4°C (up to 2 years); in solvent at -80°C (up to 6 months).
  • Deuterium-labeled internal standard for quantitative NMR, GC-MS, and LC-MS.
  • High purity (98.0%) suitable for analytical workflows.
  • Solid powder form with defined long-term storage conditions.
  • Provides molecular weight and structural information for method development.
  • Available in small pack sizes for tracer and reference applications.
Catalog No. 50-002-97090 Supplier Medchemexpress LLC Supplier No. HYB0631S10MG
Alendronic acid-d6 is the deuterium-labeled analogue of alendronic acid supplied as a solid stable-isotope reagent for use as a tracer and internal standard in quantitative analytical methods such as NMR, GC-MS, and LC-MS. Purity 98.0%; molecular weight 255.13; molecular formula C4H7D6NO7P2. Storage: powder at -20°C (up to 3 years) or 4°C (up to 2 years); in solvent at -80°C (up to 6 months).
  • Deuterium-labeled internal standard for quantitative NMR, GC-MS, and LC-MS.
  • High purity (98.0%) suitable for analytical workflows.
  • Solid powder form with defined long-term storage conditions.
  • Provides molecular weight and structural information for method development.
  • Available in small pack sizes for tracer and reference applications.