Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abbott™ Acceava™ Strep A Test

Catalog No. 23875550c Shop All Abbott Rapid Diagnostics Products
Change view
Click to view available options
Quantity:
Each
Catalog No. Quantity
23875550C Each
1 options
This item is not returnable. View return policy

Catalog No. 23875550C

Supplier: Abbott™ 4580295008

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

High-performance, easy-to-use Strep A test for use in CLIA-waived laboratories

  • Sensitivity: 97.0%
  • Specificity: 95.0%
  • Overall accuracy: 95%
  • Positive results in as little as one minute
  • One-minute hands-on time
  • Reimbursable

Specifications

Clia Complexity Waived
Detectable Analytes Streptococcal Antigen (Group A)
Type Streptococcus A Test
Includes Test strip
Quantity Each
Test Time 1 min.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.