Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
Includes
Test devices, transfer pipettes, positive and negative controls (CLIA Waived tests also include sterile NP swabs)
- CLIA Complexity: Waived and Moderate (see table)
- CPT Code: 87804QW and 87804 (see table)
- Sensitivity: 100% (Flu A); 93% (Flu B)
- Specificity: 96% (Flu A); 97% (Flu B)
- One-step, rapid immunochromatographic assay
- Qualitative detection of influenza A and B nucleoprotein antigens
- Avian influenza H5N1 isolates included in list of influenza strains
- Results within 15 minutes†
- Specimens: Nasopharyngeal (NP) swab and nasal wash/aspirate
- For in vitro diagnostic use only
Specifications
Specifications
| Clia Complexity | Moderate |
| Detectable Analytes | Influenza A and B |
| Sample Type | Nasal Swab, Nasopharyngeal Swab, Nasopharyngeal Wash/Aspirate |
| Type | Test Kit |
| Includes | 22 Test Units, transfer pipettes, positive control swab (Inactivated Influenza A and B), negative control swab (Inactivated Strep A), elution vials for control, product insert and procedure card |
| Quantity | 22 Tests |
| Test Time | 12 min. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?