Learn More
Abbott™ BinaxNOW™ Nasopharyngeal Swab Accessory Pack
GSA_VA
Catalog No. BNX400065 Shop All Abbott Rapid Diagnostics Products
Change view
Click to view available options
Quantity:
Each
Catalog No. Quantity
BNX400065 Each
1 options
This item is not returnable. View return policy
Catalog No. BNX400065 Supplier Abbott™ Supplier No. 400065
Add to Cart
Add to Cart
This item is not returnable. View return policy
Swabs and transport vials for use with BinaxNOW™ Diagnostic test kits.
Includes:
  • 20 Sterile foam swabs
  • Elution solution transport vials
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Type Nasopharyngeal Swab Accessory Pack
Includes 20 Sterile Foam swabs and Elution solution transport vial
Quantity Each