Learn More
Description
Includes
Test devices, pipettes, control swabs and control reagents
Immunochromatographic assay for the qualitative detection of RSV fusion protein antigen in neonatal and pediatric patients under the age of five
- Time for test results: 15 minutes
- Sample type: Nasal wash and nasopharyngeal (NP) swabs
- Nasal Wash: Sensitivity 89%, Specificity 100%
- NP Swabs: Sensitivity 93%, Specificity 93%
- Intended for in vitro diagnostic use
Kit BNX-430000:
- CLIA Complexity: Moderate
- CPT Code: 87807
- Does not include NP specimen swabs or NP Swab Accessory Pack (BNX-400065)
- CLIA Complexity: Waived
- CPT Code: 87807QW
- Include NP specimen swabs and NP Swab Accessory Pack (BNX-400065)
Specifications
Specifications
| CPT Code | 87807 |
| Clia Complexity | Moderate |
| Detectable Analytes | Respiratory Syncytial Virus (RSV) |
| Format | Patented Card |
| Sample Type | Nasal wash and nasopharyngeal swab |
| Type | RSV Test Kit |
| Includes | 42 test cards, transfer pipettes, control swabs and control reagents, product insert and procedural card |
| Quantity | 42 Tests |
| Test Time | 15 min. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.