Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abbott™ BinaxNOW™ RSV Lateral Flow Test Kits

Catalog No. BNX430100 Shop All Abbott Rapid Diagnostics Products
Change view
Click to view available options
Clia Complexity:
Moderate
Waived
CPT Code:
87807
87807QW
Quantity:
10 Tests
22 Tests
42 Tests
Includes:
10 test cards, transfer pipettes, control swabs and control reagents, transport vials, collection swabs and NP swab accessory pack (BNX-400065), product insert and procedural card
42 test cards, transfer pipettes, control swabs and control reagents, product insert and procedural card
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Clia Complexity CPT Code Quantity Includes
BNX430100 Waived 87807QW 10 Tests 10 test cards, transfer pipettes, control swabs and control reagents, transport vials, collection swabs and NP swab accessory pack (BNX-400065), product insert and procedural card
BNX430122 Waived 87807QW 22 Tests 10 test cards, transfer pipettes, control swabs and control reagents, transport vials, collection swabs and NP swab accessory pack (BNX-400065), product insert and procedural card
BNX430000 Moderate 87807 42 Tests 42 test cards, transfer pipettes, control swabs and control reagents, product insert and procedural card
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. BNX430100 Supplier Abbott™ Supplier No. 430100
Only null left
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Includes

Test devices, pipettes, control swabs and control reagents

Rapid, one-step lateral flow test for Respiratory Syncytial Virus (RSV)

Immunochromatographic assay for the qualitative detection of RSV fusion protein antigen in neonatal and pediatric patients under the age of five

  • Time for test results: 15 minutes
  • Sample type: Nasal wash and nasopharyngeal (NP) swabs
  • Nasal Wash: Sensitivity 89%, Specificity 100%
  • NP Swabs: Sensitivity 93%, Specificity 93%
  • Intended for in vitro diagnostic use

Kit BNX-430000:

  • CLIA Complexity: Moderate
  • CPT Code: 87807
  • Does not include NP specimen swabs or NP Swab Accessory Pack (BNX-400065)
Kits BNX-430100 and BNX-430122:
  • CLIA Complexity: Waived
  • CPT Code: 87807QW
  • Include NP specimen swabs and NP Swab Accessory Pack (BNX-400065)

Specifications

CPT Code 87807QW
Clia Complexity Waived
Detectable Analytes Respiratory Syncytial Virus (RSV)
Format Patented Card
Sample Type Nasal wash and nasopharyngeal swab
Type RSV Test Kit
Includes 10 test cards, transfer pipettes, control swabs and control reagents, transport vials, collection swabs and NP swab accessory pack (BNX-400065), product insert and procedural card
Quantity 10 Tests
Test Time 15 min.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.