Learn More
Abbott™ BioStar™ Acceava™ hCG Control Kit
Catalog No. 23875595 Shop All Abbott Rapid Diagnostics Products
Change view
Click to view available options
Quantity:
Each
Catalog No. Quantity
23-875-595 Each
1 options
This item is not returnable. View return policy
Exact match for part number alr92218
Catalog No. 23-875-595 Supplier Abbott™ Supplier No. 92218
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Exact match for part number alr92218
Control Kit for use with Alere BioStar Acceava and Clearview hCG Tests
  • Use with several types of hCG tests to ensure accuracy
  • Includes one positive and one negative control
Specifications
Content And Storage 15°C to 25°C
Type BioStar Acceava hCG Control Kit
Includes 1 Positive and 1 negative control
Intended Use Alere BioStar acceava and clearview hCG test
Quantity Each