Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abbott™ RS-232 Adapter Kit

Catalog No. 23762970 Shop All Abbott Rapid Diagnostics Products
Change view
Click to view available options
Content And Storage:
15°C to 30°C (59°F to 86°F)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Content And Storage
23-762-970 15°C to 30°C (59°F to 86°F)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-762-970 Supplier Abbott™ Supplier No. 10958
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Allows interface between the Cholestech LDX™ System and PC-Compatible Computers for patient result download.

  • Allows interface between the LDX System and PC-Compatible Computers for patient result download

Specifications

Type RS232 Adaptor Kit
For Use With (Application) Allows interface between the LDX System and PC-Compatible Computers for patient result download
For Use With (Equipment) Alere Cholestech LDX analyzer and PC-compatible computers
Includes LDX RS232 adapter kit and package insert
Content And Storage 15°C to 30°C (59°F to 86°F)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.