Learn More
Abbott™ Clearview™ hCG Tests
Catalog No. 23046595 Shop All Abbott Rapid Diagnostics Products
Change view
Click to view available options
Clia Complexity:
Waived
Waived (Urine), Non-waived (Serum)
Detectable Analytes:
hCG
Quantity:
40 Tests
50 Tests
Type:
25 hCG Combo Cassette
hCG Combo II Cassette
hCG Urine Cassettes
hCG Urine Dipstick
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Clia Complexity Detectable Analytes Quantity Type
23-046-595 Waived (Urine), Non-waived (Serum) hCG 40 Tests hCG Combo II Cassette
23-046-596 Waived hCG 40 Tests hCG Urine Cassettes
23-046-597 Waived hCG 50 Tests hCG Urine Dipstick
23-046-613 Waived (Urine), Non-waived (Serum) hCG 40 Tests 25 hCG Combo Cassette
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 23-046-595 Supplier Abbott™ Supplier No. 92215
Add to Cart
Add to Cart
Innovative one-step tests provide highly sensitive and specific results for the early detection of the human pregnancy hormone hCG
  • CLIA Complexity: Waived for urine, Non-Waived for serum
  • Variety of easy-to-use, single-step tests
  • Highly sensitive assay with built-in controls ensures accuracy
  • Test results in under five minutes
Specifications
Clia Complexity Waived (Urine), Non-waived (Serum)
Detectable Analytes hCG
Sample Type Serum, Urine
Type hCG Combo II Cassette
Quantity 40 Tests
Test Time 5 min. or less