Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abbott™ Clearview™ Mono Test

Catalog No. 23046598 Shop All Abbott Rapid Diagnostics Products
Change view
Click to view available options
Quantity:
30 Cassettes
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-046-598 30 Cassettes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-046-598 Supplier Abbott™ Supplier No. 4581515020

missing translation for 'validMainframeMsg'

Provides highly sensitive and specific tests for infectious mononucleosis

  • CLIA Complexity: Waived for whole blood, Non-Waived for serum or plasma
  • Sensitivity: 98%
  • Specificity: >99%
  • Use with multiple sample types — whole blood, serum or plasma
  • Easy to use two-step test requires no training
  • Built-in controls ensure accuracy

Specifications

Clia Complexity Waived, Non-waived
Format Cassette
Type Mononucleosis Test Kit
Quantity 30 Cassettes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.