Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abbott™ Clearview™ Strep A Exact II Test Kits

Catalog No. 23046599 Shop All Abbott Rapid Diagnostics Products
Change view
Click to view available options
Quantity:
30 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-046-599 30 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-046-599 Supplier Abbott™ Supplier No. 4581225020
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Provide highly sensitive and specific test for Group A Streptococcal Antigen

  • Sensitivity: 97%
  • Specificity: 95%
  • Direct detection of Group A Strep from throat swabs
  • Built-in control ensures accuracy
  • Test results in five minutes or less

Specifications

Clia Complexity Waived
Detectable Analytes Streptococci (Group A)
Sample Type Throat Swab
Type Streptococcus A Dipstick Test Kit
Quantity 30 Tests
Test Time 1 min.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.