Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abbott™ Determine™ HIV-1/2 Ag/Ab Combo, External Controls Kit
GSA_VA

Catalog No. 23046615 Shop All Abbott Rapid Diagnostics Products
Change view
Click to view available options
Quantity:
4 Vials
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-046-615 4 Vials
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-046-615 Supplier Abbott™ Supplier No. 7D2628
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Determine™ HIV External Controls Kit

  • Contains 4, 1.5 mL vials, each allowing 30 testing events
  • 1 vial each of: HIV Non-Reactive, HIV-1 p24 positive, HIV-1 positive, and HIV-2 positive

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Sample Type Plasma, Serum, Venous Whole Blood
Type HIV Testing External Control
Quantity 4 Vials
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.