Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GIARDIA/CRYPTOSPORIDIUM QUIK CHEK™
GSA_VA

Catalog No. 23046789
Change view
Click to view available options
Quantity:
25 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-046-789 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-046-789 Supplier Techlab Inc Supplier No. T30407
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Eliminates complicated, labor-intensive processes while maintaining high levels of accuracy. Simultaneously detects and differentiates Giardia cyst antigen and Cryptosporidium oocyst antigen. Developed and Manufactured by TECHLAB, Inc.

  • Results in 30 minutes without instrumentation
  • Eliminates subjective interpretation of results
  • Giardia: Sensitivity 98.9% / Specificity 100%
  • Cryptosporidium: Sensitivity 100% / Specificity 99.8%
  • CLIA: Moderate Complexity

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

CPT Code 87328, 87329
Clia Complexity Moderate
Detectable Analytes Giardia Antigen, Cryptosporidium Antigen
Type Giardia/Cryptosporidium QUIK CHEK™
Includes 25 Membrane device pouches, diluent, wash buffer, conjugate, substrate, positive control, disposable plastic transfer pipettes
Quantity 25 Tests
Test Time 30 min.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.