Learn More
Techlab C. DIFF QUIK CHEK COMPLETE™
Description
- Results in approximately 30 minutes
- Easy to intrepret
- Highly sensitive and specific
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Detectable Analytes | C. difficile GDH Antigen, Toxins A and B |
| Type | Clostridium difficile QUIK CHEK COMPLETE™ |
| Includes | 25 Membrane device pouches, diluent, wash buffer, conjugate, substrate, positive control, disposable plastic transfer pipettes |
| Quantity | 25 Tests |