Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abbott™ ID NOW Strep A 2 Control Swab Kit

Catalog No. 23046633 Shop All Abbott Rapid Diagnostics Products
Change view
Click to view available options
Quantity:
12 Sets
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-046-633 12 Sets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-046-633 Supplier Abbott™ Supplier No. 734080
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

12 Positive and 12 Negative Swabs

Abbott ID NOW Strep A 2 Positive and Negative Control Swabs. May be used to demonstrate the ability to generate appropriate positive and negative results by following the assay process. Refer to PI for frequency.

Good laboratory practice suggests the use of positive and negative controls to ensure that test reagents are working and that the test is correctly performed. The Abbott™ ID NOW Strep A 2 kit contains Positive and Negative Control Swabs. These swabs may be used to demonstrate the ability to generate appropriate positive and negative results by following the assay process. Test these swabs once when the assay is run on an instrument for the first time, as well as with each new shipment received and once for each untrained operator. Further controls may be tested in order to conform with local, state and/or federal regulations, accrediting groups, or your lab’s standard Quality Control procedures.

Specifications

Content And Storage 2°C to 30°C
For Use With (Application) For accurate detection of Group A Strep
Includes Abbott ID NOW STREP A 2 Positive and Negative Control Swabs (12 Positive and 12 Negative Swabs)
Quantity 12 Sets
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.