Learn More
Abbott™ iCassette™ Single Test Drug Screen Cassettes
Catalog No. 22911074
Change view
Click to view available options
Quantity:
40/Pk.
Catalog No. Quantity
22-911-074 40/Pk.
1 options
This item is not returnable. View return policy
Catalog No. 22-911-074 Supplier Abbott™ Supplier No. DBU102
Add to Cart
Add to Cart
This item is not returnable. View return policy
Rapid and accurate screening of drugs in minutes
  • Available as single drug testing cassette
  • Requires small amount of urine (3mL) to run
  • Pipet included for procedure testing
  • Administrator remains in control of test device
Specifications
Type Buprenorphine (BUP) Single Test Cassette
For Use With (Application) For screening single drug in urine sample
Quantity 40/Pk.