Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abbott™ IMPACT RPR™ Test Cards

Catalog No. p-6830105 Shop All Abbott Rapid Diagnostics Products
Click to view available options
Quantity:
300 Pack
Type:
10-well Test Cards

IMPACT RPR™ Test Cards

  • Classified as a standard test for syphilis by the Centers for Disease Control and Prevention (CDC)
  • Non-treponemal test for the serological detection of syphilis
  • Assay utilizes charcoal particle technology

RPR test cards, 18mm circle, 10-well, 300/pk

Specifications

Type 10-well Test Cards
For Use With (Application) IMPACT RPR™
Product Line IMPACT RPR™
Quantity 300 Pack
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.