Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abbott™ IMPACT RPR™ Test Cards

Catalog No. wa61c5 Shop All Abbott Rapid Diagnostics Products
Change view
Click to view available options
Quantity:
300 Pack
Type:
10-well Test Cards
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Type
WA-61C5 300 Pack 10-well Test Cards
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. WA-61C5 Supplier Abbott™ Supplier No. 61C5
Only null left
Add to Cart
Add to Cart

IMPACT RPR™ Test Cards

  • Classified as a standard test for syphilis by the Centers for Disease Control and Prevention (CDC)
  • Non-treponemal test for the serological detection of syphilis
  • Assay utilizes charcoal particle technology

RPR test cards, 18mm circle, 10-well, 300/pk

Specifications

Type 10-well Test Cards
For Use With (Application) IMPACT RPR™
Product Line IMPACT RPR
Quantity 300 Pack
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.