Learn More
Abbott™ White Top Cup with Temperature Strip
Catalog No. 22911043
Click to view available options
Quantity:
25 Boxes
Catalog No. 22-911-043
Supplier: Abbott™ 190058
Urine collection cup helps to ensure specimen freshness and maintain chains of custody
  • Sterile 3 oz. (90mL) clear plastic cup with white cap
  • Includes temperature strip to ensure sample freshness
  • Each cup is easy to use and includes a security seal and specimen information label
Specifications
For Use With (Application) Urinalysis
Quantity 25 Boxes
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.