Learn More
Abbott™ iCassette™ Dx Urine Drug Screen
Catalog No. 22911027
Change view
Click to view available options
Quantity:
25 Pack
Catalog No. Quantity
22-911-027 25 Pack
1 options
This item is not returnable. View return policy
Catalog No. 22-911-027 Supplier Abbott™ Supplier No. IDCB1115011
Add to Cart
Add to Cart
This item is not returnable. View return policy
Rapidly screen for 11 illicit and prescription drugs
The iCassette Dx is easy to use and screens for 11 drugs on one device
  • FDA 510(k) Cleared and CLIA Waived
  • Integrated positive and negative reactive controls validate device performance
  • Simple procedure: collect and pipette to test — results in as little as 5 minutes
  • Test device remains in control of administrator
  • Requires only 5mL of specimen
  • Results can be photocopied for record keeping purposes
  • 12-18 Month shelf life
  • Results are easy to read
  • Room temperature storage
Specifications
Clia Complexity Waived
Detectable Analytes Amphetamine (AMP), Barbiturates (BAR), Benzodiazepines (BZO), Cocaine (COC), Methylenedioxymethamphetamine (MDMA), Opiates (OPI), Oxycodone (OXY), Phencyclidine (PCP), Propoxyphene (PPX), Tricyclic Antidepressants (TCA), Cannabinoids (THC)
Sample Type Urine
Type iCassette™ Dx Urine Drug Screen Panel
Quantity 25 Pack
Test Time 5 min.