Learn More
Abbott™ iCup™ A.D. Zero Exposure Urine Drug Screens
Catalog No. 22911001
Change view
Click to view available options
Detectable Analytes:
Cocaine (COC), Cannabinoids (THC), Morphine (MOP), Amphetamine (AMP), Methamphetamine (mAMP), Phencyclidine (PCP), Benzodiazepines (BZO), Oxycodone (OXY)
Cocaine (COC), Cannabinoids (THC), Opiates (OPI), Amphetamine (AMP), Methamphetamine (mAMP)
Cocaine (COC), Cannabinoids (THC), Opiates (OPI), Amphetamine (AMP), Methamphetamine (mAMP), Benzodiazepines (BZO)
Cocaine (COC), Cannabinoids (THC), Opiates (OPI), Amphetamine (AMP), Methamphetamine (mAMP), Benzodiazepines (BZO), Barbiturates (BAR), Oxycodone (OXY), Methodone (MTD), Propoxyphene (PPX)
Cocaine (COC), Cannabinoids (THC), Opiates (OPI), Amphetamine (AMP), Methamphetamine (mAMP), Phencyclidine (PCP)
Cocaine (COC), Cannabinoids (THC), Opiates (OPI), Amphetamine (AMP), Methamphetamine (mAMP), Phencyclidine (PCP), Benzodiazepines (BZO), Barbiturates (BAR)
Cocaine (COC), Cannabinoids (THC), Opiates (OPI), Amphetamine (AMP), Methamphetamine (mAMP), Phencyclidine (PCP), Benzodiazepines (BZO), Barbiturates (BAR), Methodone (MTD)
Cocaine (COC), Cannabinoids (THC), Opiates (OPI), Amphetamine (AMP), Methamphetamine (mAMP), Phencyclidine (PCP), Benzodiazepines (BZO), Barbiturates (BAR), Methodone (MTD), Tricyclic Antidepressants (TCA)
Cocaine (COC), Cannabinoids (THC), Opiates (OPI), Amphetamine (AMP), Methamphetamine (mAMP), Phencyclidine (PCP), Benzodiazepines (BZO), Barbiturates (BAR), Methodone (MTD), Tricyclic Antidepressants (TCA), Oxycodone (OXY), Propoxyphene (PPX)
Cocaine (COC), Cannabinoids (THC), Opiates (OPI), Amphetamine (AMP), Phencyclidine (PCP)
Cocaine (COC), Cannabinoids (THC), Opiates (OPI), Methamphetamine (mAMP)
Cocaine (COC), Cannabinoids (THC), Opiates (OPI), Methamphetamine (mAMP), Phencyclidine (PCP)
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Detectable Analytes
22-911-001 Cocaine (COC), Cannabinoids (THC), Opiates (OPI), Methamphetamine (mAMP)
22-911-002 Cocaine (COC), Cannabinoids (THC), Opiates (OPI), Amphetamine (AMP), Methamphetamine (mAMP)
22-911-005 Cocaine (COC), Cannabinoids (THC), Opiates (OPI), Amphetamine (AMP), Methamphetamine (mAMP), Phencyclidine (PCP)
22-911-003 Cocaine (COC), Cannabinoids (THC), Opiates (OPI), Amphetamine (AMP), Phencyclidine (PCP)
22-911-004 Cocaine (COC), Cannabinoids (THC), Opiates (OPI), Methamphetamine (mAMP), Phencyclidine (PCP)
22-911-006 Cocaine (COC), Cannabinoids (THC), Opiates (OPI), Amphetamine (AMP), Methamphetamine (mAMP), Benzodiazepines (BZO)
22-911-007 Cocaine (COC), Cannabinoids (THC), Opiates (OPI), Amphetamine (AMP), Methamphetamine (mAMP), Phencyclidine (PCP), Benzodiazepines (BZO), Barbiturates (BAR)
22-911-009 Cocaine (COC), Cannabinoids (THC), Opiates (OPI), Amphetamine (AMP), Methamphetamine (mAMP), Phencyclidine (PCP), Benzodiazepines (BZO), Barbiturates (BAR), Methodone (MTD)
22-911-011 Cocaine (COC), Cannabinoids (THC), Opiates (OPI), Amphetamine (AMP), Methamphetamine (mAMP), Benzodiazepines (BZO), Barbiturates (BAR), Oxycodone (OXY), Methodone (MTD), Propoxyphene (PPX)
22-911-008 Cocaine (COC), Cannabinoids (THC), Morphine (MOP), Amphetamine (AMP), Methamphetamine (mAMP), Phencyclidine (PCP), Benzodiazepines (BZO), Oxycodone (OXY)
22-911-012 Cocaine (COC), Cannabinoids (THC), Opiates (OPI), Amphetamine (AMP), Methamphetamine (mAMP), Phencyclidine (PCP), Benzodiazepines (BZO), Barbiturates (BAR), Methodone (MTD), Tricyclic Antidepressants (TCA), Oxycodone (OXY), Propoxyphene (PPX)
22-911-010 Cocaine (COC), Cannabinoids (THC), Opiates (OPI), Amphetamine (AMP), Methamphetamine (mAMP), Phencyclidine (PCP), Benzodiazepines (BZO), Barbiturates (BAR), Methodone (MTD), Tricyclic Antidepressants (TCA)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
This item is not returnable. View return policy
Catalog No. 22-911-001 Supplier Abbott™ Supplier No. IDUA147012
Add to Cart
Add to Cart
This item is not returnable. View return policy
A zero exposure urine drug screen that tests for up to 12 drugs at once
Detect the presence of drugs and adulterants in just minutes
  • The iCup AD Drug Screen offers versatility with up to 12 drug assays in one cup
  • Multiple configurations available
  • Built-in validity screening to alleviate tampering concerns
  • Room temperature storage
  • Comes with temperature strip to verify range of collected sample
  • Results can be photocopied for record keeping purposes
  • 24-month shelf life
Specifications
Detectable Analytes Cocaine (COC), Cannabinoids (THC), Opiates (OPI), Methamphetamine (mAMP)
Sample Type Urine
Type iCup™ A.D. Urine Drug Screen
Quantity 25 Pack