Learn More
Abbott™ iCup™ Zero Exposure Urine Drug Screen
Catalog No. 22911013
Change view
Click to view available options
Quantity:
25 Pack
Catalog No. Quantity
22-911-013 25 Pack
1 options
This item is not returnable. View return policy
Catalog No. 22-911-013 Supplier Abbott™ Supplier No. IDOA1137011
Add to Cart
Add to Cart
This item is not returnable. View return policy
A zero exposure urine drug screen that tests for up to 13 drugs in one device
Detect the presence of drugs in just minutes
  • The iCup Drug Screen offers versatility through screening for 13 different drug assays in one cup
  • Room temperature storage
  • Comes with temperature strip to verify specimen validity
  • Results can be photocopied for record keeping purposes
Specifications
Detectable Analytes Cocaine (COC), Cannabinoids (THC), Opiates (OPI), Amphetamine (AMP), Methamphetamine (mAMP), Phencyclidine (PCP), Benzodiazepines (BZO), Barbiturates (BAR), Methadone (MTD), Tricyclic Antidepressants (TCA), Oxycodone (OXY), Propoxyphene (PPX), Buprenorphine (BUP)
Sample Type Urine
Type iCup™ Zero Exposure Urine Drug Screen Panel
Quantity 25 Pack
Test Time 5 min.