Learn More
Abbott™ Instant Bluing Tablets
Catalog No. 22911042
Change view
Click to view available options
Quantity:
100 Tablets
Catalog No. Quantity
22-911-042 100 Tablets
1 options
Catalog No. 22-911-042 Supplier Abbott™ Supplier No. 7875
Add to Cart
Edge
Add to Cart
Prevent adulteration prior to specimen collection
  • Instant bluing tablets — designed for quick, portable and easy-to-use bluing application
  • Used to protect the integrity of urine samples by coloring toilet water, which can be intentionally used to dilute a urine sample
  • Packaged in a twist cap bottle; 100 tablets per bottle
Specifications
Type Instant Bluing Tablet
For Use With (Application) Prevent adulteration prior to specimen collection
Quantity 100 Tablets