Learn More
Abbott™ Integrated E-Z Key Cup 12-Panel Drug Screen
Catalog No. 22911017
Change view
Click to view available options
Quantity:
25 Pack
Catalog No. Quantity
22-911-017 25 Pack
1 options
This item is not returnable. View return policy
Catalog No. 22-911-017 Supplier Abbott™ Supplier No. DOA1127041019
Add to Cart
Add to Cart
This item is not returnable. View return policy
A split specimen urine drug screen that tests for up to 12 drugs in one device
Detect the presence of drugs in just minutes
  • All-inclusive drug testing device for split specimen collection; isolates specimen portions, with one for the instant result test and one untouched for laboratory confirmation testing results, minimizing the risk of inconclusive confirmation testing results
  • Comes with temperature strip to verify specimen validity
  • Self-contained cup eliminates collector exposure
  • Results can be photocopied for record keeping purposes
  • 12-18 Month shelf life
  • Room temperature storage
Specifications
Detectable Analytes Cocaine (COC), Cannabinoids (THC), Morphine (MOP), Amphetamine (AMP), Methamphetamine (mAMP), Benzodiazepines (BZO), Barbiturates (BAR), Methadone (MTD), Methylenedioxymethamphetamine (MDMA), Oxycodone (OXY), Propoxyphene (PPX), Buprenorphine (BUP)
Sample Type Urine
Type E-Z Key Cup Drug Screen Panel
Quantity 25 Pack
Test Time Results in minutes