Learn More
Abbott™ iScreen™ Urine Drug Control Kit
Description
- The external quality controls are designed to work with any Alere urine drug screens
- Includes a positive and negative vial, 5mL each
- High positive is 3X cutoff
- Refrigeration required
Specifications
Specifications
| Sample Type | Urine |
| Type | iScreen Urine Drug Control Kit |
| Includes | Positive, negative vial 5mL |
| Intended Use | For Urine drug screens, validation of test performance |
| Quantity | 2 Boxes |