Learn More
Abbott™ NicQuick™ Nicotine/Cotinine Test
Catalog No. 22911044
Change view
Click to view available options
Quantity:
40/Pk.
Catalog No. Quantity
22-911-044 40/Pk.
1 options
Catalog No. 22-911-044 Supplier Abbott™ Supplier No. DCT102
Add to Cart
Add to Cart
For the determination of smoking status
  • NicQuick detects the presence of continine in urine at the cut-off concentration of 200ng/mL
  • Cotinine, a nicotine metabolite, is detectable in urine up to three days after smoking
Specifications
Detectable Analytes Nicotine, Cotinine
Sample Type Urine
Type Nicotine/Cotinine Urine Test
Quantity 40/Pk.