Learn More
Abbott™ iScreen™ Saliva Alcohol Test Strips
Catalog No. 22911076
Change view
Click to view available options
Quantity:
25/Pk.
Catalog No. Quantity
22-911-076 25/Pk.
1 options
This item is not returnable. View return policy
Catalog No. 22-911-076 Supplier Abbott™ Supplier No. ISA25
This item is not returnable. View return policy
Convenient and rapid preliminary test that detects saliva alcohol concentration (SAC)
Specifications
Type iScreen Saliva Alcohol Test Strips
For Use With (Application) Detects saliva alcohol concentration (0.0%, 0.02%, 0.08% and 0.30% SAC)
Quantity 25/Pk.