Learn More
Abbott™ PRISM (Precalibrated Replaceable Intelligent Sensor Module) for AlcoMate™ Premium AL7000
Catalog No. 22911080
Change view
Click to view available options
For Use With (Equipment):
AlcoMate Premium Model AL7000 Breathalyzer
Catalog No. For Use With (Equipment)
22-911-080 AlcoMate Premium Model AL7000 Breathalyzer
1 options
Catalog No. 22-911-080 Supplier Abbott™ Supplier No. PRISM
Add to Cart
Add to Cart
For use with AlcoMate Premium Model AL7000 Breathalyzer.
  • Dimensions: 1.27 x 1.27 x 1.9cm
Note
Replace the sensor module every 200 to 300 tests, or at least once yearly.
Order Info
Replace the sensor module every 200 to 300 tests, or at least once yearly.
Specifications
Type PRISM (Precalibrated Replaceable Intelligent Sensor Module)
For Use With (Equipment) AlcoMate Premium Model AL7000 Breathalyzer