Learn More
SHIGA TOXIN QUIK CHEK™
GSA_VA
Catalog No. 23900553
Change view
Click to view available options
Quantity:
25 Tests
Catalog No. Quantity
23-900-553 25 Tests
1 options
This item is not returnable. View return policy
Catalog No. 23-900-553 Supplier Techlab Inc Supplier No. T30625
 Restricted Item
Add to Cart
Add to Cart
This item is not returnable. View return policy
Aids in diagnosis of disease caused by Shiga toxin-producing Escherichia coli (STEC) hours earlier than many other tests. Rapid, qualitative assay detects and differentiates STEC toxins Stx1 – and Stx2 – directly from fecal samples. Developed and Manufactured by TECHLAB, Inc.
  • Offers analytical sensitivity of 0.04ng/mL
  • Uses TECHLAB QUIK CHEK™ test format
  • Provides results in less than 30 minutes
  • Diagnosis up to 24 hours earlier than other rapid tests or traditional methods
  • Compatible with primary fecal samples, broth, plate cultures and specimens in transport media
Test Principle
  • Employs rapid membrane enzyme immunoassay
  • Membrane Device contains reaction Window with two vertical lines of immobilized antibodies for Stx1 and Stx2, plus anti-horseradish peroxidase (HRP) control line
  • Sample is mixed with diluent and conjugate and applied to sample well
  • Stx1 and/or Stx2 in sample bind to antibody-peroxidase conjugates and are subsequently captured by immobilized Stx1 and Stx2 specific monoclonal antibodies in test lines
  • Positive control reaction is indicated by vertical dotted blue line
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Detectable Analytes E. coli Shiga Toxin 1 and 2
Type Shiga Toxin QUIK CHEK™
Includes 25 Membrane device pouches, diluent, wash buffer, conjugate, substrate, positive control, disposable plastic transfer pipettes
Quantity 25 Tests
Test Time 30 min.