Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TOX A/B QUIK CHEK™
GSA_VA

Catalog No. 22293515
Change view
Click to view available options
Quantity:
25 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-293-515 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-293-515 Supplier Techlab Inc Supplier No. 30394
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

25 Membrane device pouches, each containing a device and dessicant; Diluent (14mL); Wash buffer (10mL); Conjugate (2mL); Substrate (3.5mL); Positive control (1mL); Disposable plastic transfer pipets (50)

Rapid enzyme immunoassay cassette for detecting C. difficile toxins A and B in fecal specimens. Specimens can be run individually or batched. Developed and Manufactured by TECHLAB, Inc.

  • Larger sample well makes testing fast, easy, accurate and clean
  • Unique technology smoothly disperses sample within the device
  • Chance of clogging is minimized, reducing spillovers
  • Easy to read — two lines mean C. difficile toxin A/B is present
  • Internal control line validates the QC result in the same reaction window
  • Sensitivity: ≥90%
  • Specificity: ≥99%

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Detectable Analytes C. difficile Toxins A and B
Type Clostridium difficile QUIK CHEK™
Includes 25 Membrane device pouches, diluent, wash buffer, conjugate, substrate, positive control, disposable plastic transfer pipettes
Quantity 25 Tests
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.