Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abbott™ Clearview™ RSV Test Kit
GSA_VA

Catalog No. wa135060 Shop All Abbott Rapid Diagnostics Products
Change view
Click to view available options
Quantity:
20 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
WA135060 20 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. WA135060 Supplier Abbott™ Supplier No. 135060
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Includes

Test devices (20), extraction reagent solution (2mL), extraction tubes (20), disposable pipettes (40), positive control (1mL), negative control (1mL)

Intended for the rapid, qualitative detection of respiratory syncytial virus (RSV) directly from nasopharyngeal swab and nasal aspirate specimens in those under the age of 6 or over the age of 60. Intended for use as an aid in the diagnosis of patients with symptoms consistent with RSV infection.

 

  • Sensitivity: 93.8%
  • Specificity: 97.7%
  • Built-in control ensures accuracy
  • For use with nasopharyngeal swabs and nasal aspirate specimens
  • Approved claim: in children less than six years and adults over the age of 60

 

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Certifications/Compliance CE Marked
CPT Code 87899
Clia Complexity Waived
Detectable Analytes Respiratory Syncytial Virus
Sample Type Nasopharyngeal swab or aspirates
Type RSV Test Kit
Includes 20 Test devices, 2mL extraction reagent solution, 20 extraction tubes, 40 disposable pipettes, 1mL positive control, 1mL negative control, 1 package Insert
Quantity 20 Tests
Test Time 15 min.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.