Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abbott™ DSX™/DS2™ Automated ELISA Processing System Consumables - Dilution Wells

Catalog No. 23046923 Shop All Abbott Rapid Diagnostics Products
Change view
Click to view available options
Product Type:
Dilution Strips, 8-well
Macrowell Tube Strips
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
23-046-923 Macrowell Tube Strips
23-046-890 Dilution Strips, 8-well
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23-046-923 Supplier Abbott™ Supplier No. 65960
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Designed for use with the Dynex™ DSX™ and DS2™ fully automated ELISA processing systems.

Designed for use with the Dynex&trade DSX&trade with a the Macrowell tube holder. Use with part # 65967.

Specifications

Depth (English) 0.18 in.
Product Type Macrowell Tube Strips
For Use With (Application) Designed for use with the Dynex™ DSX™ with a Macrowell tube holder.
Depth (Metric) 0.5 cm
Height (English) 7.1 in.
Height (Metric) 18 cm
Width (English) 3 in.
Width (Metric) 7.6 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.