Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abbott™ DS2™ Automated ELISA Processing System Consumables:15mL Reagent Tubes
GSA_VA

Catalog No. 23046922 Shop All Abbott Rapid Diagnostics Products
Change view
Click to view available options
Volume (Metric) Sample:
15 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Sample
23-046-922 15 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-046-922 Supplier Abbott™ Supplier No. 62930

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

For use with the Dynex™ DS2™ fully automated ELISA processing system.

15 mL reagent tubes with screw cap; 50/pack

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Volume (Metric) Sample 15 mL
For Use With (Application) Dynex DS2™ fully automated ELISA processing system
Product Type Reagent Tube
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.