Learn More
C. DIFFICILE TOX A/B II™
GSA_VA
Catalog No. WA30397
Change view
Click to view available options
Quantity:
96 Tests
Catalog No. Quantity
WA-30397 96 Tests
1 options
This item is not returnable. View return policy
The products indicated contain mercury. Do not put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by local, state or federal laws. Some products containing mercury are not available for sale in some states.
Catalog No. WA-30397 Supplier Techlab Inc Supplier No. 30397
Add to Cart
Add to Cart
This item is not returnable. View return policy
The products indicated contain mercury. Do not put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by local, state or federal laws. Some products containing mercury are not available for sale in some states.
Includes
12 x 8-well strips with breakaway wells, sample diluent, controls and calibrators, conjugate, TMB substrate and wash buffer
C. DIFFICILE TOX A/B II is an ELISA test for the detection of Toxins A and B in stool specimen. Rapid protocol allows for results in as little as 30 minutes. Developed and Manufactured by TECHLAB, Inc.
  • Excellent correlation to tissue culture
  • Assay performed in approximately 30 minutes
  • No gray zone eliminates equivocal results
  • Visual and instrument read options
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Detectable Analytes C. difficile Toxins A and B
Type Clostridium difficile Tox A/B Test
Includes 12 x 8-well strips with breakaway wells, sample diluent, control, calibrators, conjugate, TMB substrate, Wash Buffer
Quantity 96 Tests