Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

E. HISTOLYTICA II™
GSA_VA

Catalog No. 23046760
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-046-760 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-046-760 Supplier Techlab Inc Supplier No. 30404
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

State-of-the art ELISA test for the detection of Entamoeba histolytica pathogens in fecal specimens. Does not cross-react with E. dispar detecting only pathogenic E. histolytica. Developed and Manufactured by TECHLAB, Inc.

  • Does not cross-react with E. dispar  detecting only pathogenic E. histolytica
  • More sensitive and specific than microscopy
  • Assay can be run manually or automated
  • Break-apart microplate wells minimize reagent use
  • This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

    Specifications

    Detectable Analytes E. histolytica
    Format Microplate
    Sample Type Fecal
    Type E. histolytica II Test Kit
    For Use With (Application) For the qualitative rapid detection of Entamoeba histolytica adhesin in Human faecal specimens
    Includes Manual or automated incubation protocols, color coded and ready-to-use reagents
    Quantity 96 Tests
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.