Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abbott™ IMPACT RPR™ Liquid Controls
GSA_VA

Catalog No. WA61C3 Shop All Abbott Rapid Diagnostics Products
Change view
Click to view available options
Quantity:
3 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
WA-61C3 3 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. WA-61C3 Supplier Abbott™ Supplier No. 61C3
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

IMPACT RPR™ Liquid Controls. For use with IMPACT RPR™ test kits.

  • Classified as a standard test for syphilis by the Centers for Disease Control and Prevention (CDC)
  • Non-treponemal test for the serological detection of syphilis
  • Assay utilizes charcoal particle technology

IMPACT RPR™ Liquid Controls. Contains Liquid controls, 1 vial each, R, MR, NR. For use with IMPACT RPR™ test kits.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Type Liquid Control Test Kit
For Use With (Application) IMPACT RPR™ Test Kits
Product Line IMPACT RPR
Quantity 3 x 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.