Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abbott™ IMPACT RPR™ Pipets/Stirrers

Catalog No. WA61C4 Shop All Abbott Rapid Diagnostics Products
Change view
Click to view available options
Quantity:
500 Pack
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
WA-61C4 500 Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. WA-61C4 Supplier Abbott™ Supplier No. 61C4

missing translation for 'validMainframeMsg'

For use with the IMPACT RPR™ product line.

  • Classified as a standard test for syphilis by the Centers for Disease Control and Prevention (CDC)
  • Non-treponemal test for the serological detection of syphilis
  • Assay utilizes charcoal particle technology

Specifications

Type Pipets/Stirrers
For Use With (Application) IMPACT RPR™
Product Line IMPACT RPR
Quantity 500 Pack
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.