Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abbott™ IMPACT RPR™ Antigen Suspension

Catalog No. wa61c0 Shop All Abbott Rapid Diagnostics Products
Change view
Click to view available options
Quantity:
10 x 3mL antigen suspension
3 x 3mL antigen suspension
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
WA-61C0 3 x 3mL antigen suspension
WA-61C1 10 x 3mL antigen suspension
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. WA-61C0 Supplier Abbott™ Supplier No. 61C0
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

IMPACT RPR™ Antigen Suspension

  • Classified as a standard test for syphilis by the Centers for Disease Control and Prevention (CDC)
  • Non-treponemal test for the serological detection of syphilis
  • Assay utilizes charcoal particle technology

3 x 3mL

Specifications

For Use With (Application) For Syphilis RPR test kit
Product Line IMPACT RPR
Quantity 3 x 3mL antigen suspension
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.