Learn More
LEUKO EZ VUE™
Catalog No. 23046530
Change view
Click to view available options
Quantity:
25 Tests
Catalog No. Quantity
23-046-530 25 Tests
1 options
This item is not returnable. View return policy
Catalog No. 23-046-530 Supplier Techlab Inc Supplier No. T30355
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
For the qualitative detection of elevated levels of fecal lactoferrin, a marker for fecal leukocytes and an indicator of intestinal inflammation. A rapid cassette test that is highly standardized and reproducible. Developed and manufactured by TECHLAB, Inc.
  • Alternative to microscopy for fecal white blood cells (WBC)
  • Samples can be stored up to 2 weeks either refrigerated or at room temperature with no loss of sensitivity and can be batched or run individually for on demand testing
Specifications
Clia Complexity Moderate
Detectable Analytes Fecal Lactoferrin
Type LEUKO EZ VUE™
Includes Diluent, positive control, membrane cassette, plastic pipettes, and sample preparation devices
Quantity 25 Tests
Test Time 10 min.